missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Paper pH Strips
An easy choice for classroom experiments or demonstrations
$56.16 - $68.89
Specifications
| For Use With (Application) | For classroom experiments or demonstrations |
|---|---|
| Quantity | 100 Tests/Vial, 10 Vials/Cs. |
| Storage Requirements | Room Temperature |
| Catalog Number | Mfr. No. | pH Range | Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | pH Range | Type | Price | Quantity & Availability | ||||
|
13640509
|
Fisherbrand™
B50SHRTG030055SST |
3 to 5.5 | pH Test Paper |
Case of 10 Each for $62.23
|
|
||||
|
13640511
|
Fisherbrand™
B50SHTRG000030SST |
0 to 3 | pH Test Paper |
Case of 10 Each for $62.23
|
|
||||
|
13640512
|
Fisherbrand™
B50SHTRG080095SST |
8 to 9.5 | pH Test Paper |
Case of 10 Each for $62.23
|
|
||||
|
13640510
|
N/A | 6 to 8 | pH Test Paper |
Case of 10 Each for $67.14
|
|
||||
|
13640508
|
Fisherbrand™
B50WIDRG000140SST |
0 to 14 | pH Test Paper |
Case of 10 Each for $68.89
|
|
||||
|
13640513
|
Fisherbrand™
B50SHTRG100120SST |
10 to 12 | pH Test Paper |
Case of 10 Each for $56.16
|
|
||||
|
13640514
|
Fisherbrand™
B50SHTRG125140SST |
12.5 to 14 | pH Test Paper |
Case of 10 Each for $62.23
|
|
||||
|
13640515
|
Fisherbrand™
B50SHTRG000140SET |
0 to 14 | pH Test Paper Set |
Case of 10 Each for $62.23
|
|
||||
Description
- Ideal for educational use
- Low-cost paper strips are easy to pass around the classroom
- Easy-to-use—simply dip strip in solution to be tested and match to color chart immediately
Order Info
Box of 10 vials, 100 low-cost paper strips per vial.
Specifications
| For classroom experiments or demonstrations | |
| 100 Tests/Vial, 10 Vials/Cs. | |
| Room Temperature |