missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable HD Biohazard Waste Bags Edge
Edge

Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.

Supplier:  Fisherbrand™ 1815A

 View more versions of this product

Catalog No. 01-815A

Only null left
Edge
Explore available promotions

Description

Description

Impact- and tear-resistant autoclave bags for waste sterilization. Fisherbrand™ Orange Autoclave Bags without Sterilization Indicator are constructed of high-density polyethylene.

  • Impact- and tear-resistant.
  • Bright orange color.
  • High-density polyethylene construction

Compliance

Pass ASTM™ impact- and tear-resistance tests.

Specifications

Specifications

ASTM
Orange with Black Print
61 cm
Autoclavable
12 in.
38 micron
127°C
None
24 x 12 in.
30 cm
Printed in English
24 in.
HDPE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions