missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable Biohazard Waste Bags, with Indicator
Printed in English and Spanish.
Supplier: Fisherbrand™ 1814D
Description
Printed in English and Spanish
- Maximum temperature of 140°C (285°F); use at 125°C (257°F)
- Come with wire-tie closures
- With sterilization indicator
Specifications
| ASTM D1709, Dart impact | |
| Orange | |
| 119 cm | |
| Autoclavable | |
| 47 in. | |
| Biohazard Waste Bags with indicator |
| Wire Tie | |
| 47 x 38 in. (119 x 97 cm) | |
| 97 cm | |
| 38 in. | |
| Polypropylene | |
| 140°C |