Learn More
Fisherbrand™ Nickel Crucibles and Covers
Fisherbrand Nickel Crucibles can withstand temperatures up to 800°C (1472°F)
Description
Fisherbrand Nickel Crucibles can withstand temperatures up to 800°C (1472°F)
- Nickel Covers for all Crucibles except 13-812-131
Chemical Identifiers
| Nickel | |
| Crucible Cover |
Specifications
| Nickel | |
| For 75mL nickel crucible | |
| Crucible Cover |