missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nichrome/Aluminum Inoculating Loops

Catalog No. 131045
Change view
Click to view available options
Length (Metric):
10 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
131045 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 131045 Supplier Fisherbrand™ Supplier No. 28645
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fisherbrand Inoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. Inoculating loops are used to introduce microorganisms into a culture medium.

Fisherbrand Nichrome/Aluminum Inoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. Inoculating loops are used to introduce microorganisms into a culture medium.

  • Fisherbrand™ Nichrome/Aluminum Inoculating Loops are provided with a standard aluminum handle
  • The aluminum handle is available in either an insulated or non-insulated format
  • Inoculating loops are available in a variety of gauges, loop-hole sizes, and wire types to accommodate an array of laboratory requirements.
  • Ideal for selecting, inoculating, or transferring microbiological samples
  • Loops slide along agar surfaces without cutting or tearing material/sample
  • Cools rapidly after sterilization
  • Handle length: 8in.(203mm)
  • Passivated
  • Autoclavable up to 135°C; Can be sterilized by Eto/gamma radiation
  • Latex-free
  • Produced under ISO 9002, GMP, cGMP guidelines

Specifications

Material Nichrome wire loop; Aluminum handle
Length (English) 3.93 in.
Length (Metric) 10 cm
Sterility Sterile
Color Silver
Product Type Inoculating Loop
Gauge 25 G
Includes Tapered end aluminum handle
Length (English) Handle 8 in.
Length (Metric) Handle 20.3 cm