missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Crinkle Latex Coated Seamless Knit Cotton/Polyester Gloves

Catalog No. 19816908
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19816908 Medium
19816907 Large
19816909 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19816908 Supplier Fisherbrand™ Supplier No. 391310/M
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Ideal for assembly, construction, inspection, and general material handling.

  • Seamless cotton-polyester liner with crinkle-textured latex palm coating
  • Enhances wet and dry grip, while also repelling moisture from skin
  • Latex coating on the palm, fingertips, and thumb crotch ensures longer wear
  • Ergonomic shape reduces hand fatigue
  • Machine washable

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers, Thumb Crotch
Construction Seamless Knit
Coating Material Crinkled Latex
Color Blue, Gray
Exterior Finish Fully Textured
Grade Economy
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Medium
Style Standard
EN388 Standard 2142
For Use With (Application) Assembly, Construction, Inspection, Material Handling, and Landscaping
Glove Cuff Style Continuous Knit
Series 39-1310
Length (English) 9.7 in.
Length (Metric) 24.7 cm
Show More Show Less