missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Multi-Rod Connector

Catalog No. 02215476
Change view
Click to view available options
Size (Metric) Grip:
0 to 13 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Grip
02215476 0 to 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02215476 Supplier Fisherbrand™ Supplier No. 80862779

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Connector with side and front 13mm holes that allow creation of variety of configurations.

  • Maximum adjustment capability connector
  • Nickel-plated zinc construction
  • Rods fit into the oval hole in connector and no turning of rod occurs during tightening
  • Minimum to maximum rod size: 0 to 13mm (0 to 0.51 in.)

Specifications

Type Multi-rod Connector
Material Nickel-plated Zinc
Size (Metric) Grip 0 to 13 mm
Size (English) Grip 0 to 0.51 in.