Learn More
Fisherbrand™ Manual Differential Counter
Catalog No. 13684141
Change view
Click to view available options
No. of Keys:
5
8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Keys
13684141 8
13684140 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13684141 Supplier Fisherbrand™ Supplier No. 808CIF
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Stip chart
Designed for blood cell counting
  • Helps in automatic calculation including percentages
  • Totaling unit rings a bell at 100 increments
  • Dual reset knobs
  • Last unit is the totaling unit, which records total number of strokes made on other keys
  • Removable tabs on top of counter provide area for identification of classification being counted
  • Acid resistant, non slip bottom
Specifications
Product Type Differential Cell Counter
Displayed Text 0 to 999
No. of Keys 8
For Use With (Application) Blood Cell Counting
Includes Stip chart