missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Low-Lint Nylon Men's Inspection Gloves

Catalog No. p-143244
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19053707 Small
19053706 Medium
19013621 Large
19053708 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19053707 Supplier Fisherbrand™ Supplier No. 98740/S
Only null left

Moisture-absorbent gloves are constructed of 40-denier nylon tricot fabric and can be worn alone, or used as a glove liner.

  • 40-denier tricot fabric in a two-piece construction
  • Easy size identification by color-coded hem on rolled cuff
  • Can be worn alone or used as a liner
  • Moisture absorbent

Specifications

Construction Cut and Sewn
Coating Material Uncoated
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size Small
Sterility Sterile
For Use With (Application) Inspection, Electronics Assembly
Gender Men's
Glove Cuff Style Rolled
Hem Color Yellow
Series 98-740
Length (English) 8.7 in.
Length (Metric) 22 cm
Show More Show Less