missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Long Nose Adson Dissecting Forceps

Catalog No. 16100105
Change view
Click to view available options
Tip Style:
Long, Very Slender Nose
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
16100105 Long, Very Slender Nose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16100105 Supplier Fisherbrand™ Supplier No. PL 6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Long, slender nose for better dissecting

  • Serrations: tip and handles for better control
  • Overall length: 4.75 in., L: 4.75 in. (120mm); W: 0.384 in. (10mm) ; H: 0.73 in. (18.5mm)
  • Made of AISI 440 stainless steel

Specifications

Product Type Forceps
Material Stainless Steel
For Use With (Application) Dissection, research, laboratories, small-parts handling
Shape Straight
Tip Style Long, Very Slender Nose
Length (English) 4.75 in.
Length (Metric) 120 mm