missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lightweight Nitrile Coated Palm Gloves

Catalog No. 19816818
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19816818 Medium
19816819 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19816818 Supplier Fisherbrand™ Supplier No. 563175/M

missing translation for 'validMainframeMsg'

Thinly coated with blue nitrile on the palm and fingers to provide dexterity and breathable comfort.

  • Thinly coated with blue nitrile on the palm and fingers
  • Gloves provide chemical and cut resistance
  • Antibacterial treated cotton interlock liner resists odors and germs
  • 3 in. (8 cm) plasticized safety cuff protects wrist area
  • Ideal for automotive, assembly, fabrication, maintenance, and general handling
  • Machine washable

Specifications

Coating Material Nitrile
Color Blue
Exterior Finish Fully Textured
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size Medium
For Use With (Application) General Handling, Assembly, Fabrication, Inspection, Maintenace, and Shipping/Receiving
Glove Cuff Style Safety
Series 56-3175
Length (English) 10.6 in.
Length (Metric) 27 cm
Show More Show Less