missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Crinkle Latex Coated Cotton Jersey Gloves

Catalog No. 19816822
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19816822 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19816822 Supplier Fisherbrand™ Supplier No. 553273

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Gloves provide a sure grip while handling slippery or smooth objects.

  • Comfortable jersey liner is coated with crinkle latex
  • Ideal for general handling of glass, tile, or sheet metal
  • Gloves provide cut protection and liquid resistance
  • 3 in. (8 cm) plasticized safety cuff
  • Machine washable

Specifications

Coating Material Crinkled Latex
Color White
Exterior Finish Fully Textured
Hand Specific
Latex Free No
Material Cotton
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Glass/Tile/Sheet Metal Handling
Gender Men's
Glove Cuff Style Safety
Series 55-3273
Length (English) 15.8 in.
Length (Metric) 26 cm
Show More Show Less