missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Large Blank Writable Biohazard Identity Label
GREENER_CHOICE

Catalog No. 18999937
Change view
Click to view available options
Color:
Black, Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18999937 Black, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18999937 Supplier Fisherbrand™ Supplier No. LBH 25

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bright red fluorescent labels display the universal biohazard symbol for easy identification.

Strong adhesive ensures that labels remain in place.

Compliance

Meet OSHA requirements.

Specifications

Certifications/Compliance OSHA
Color Black, Red
Format Label
Material Paper
Product Type Biohazard Identity Label
For Use With (Application) Biohazard
Quantity 320/Pk.
Size (English) 3 x 1.93 in.
Size (Metric) 7.62 x 4.6 cm