missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lab Wipes

Catalog No. 22026345
Change view
Click to view available options
Dimensions (L x W):
12 x 13 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22026345 12 x 13 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22026345 Supplier Fisherbrand™ Supplier No. ES3900F
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highly absorbent and low-lint general purpose wipe for use throughout the clinical and research lab setting

As an alternative to paper towels and gauze, these wipes are ideal for pathology grossing and cover slipping, as well as general lab cleaning.

 

  • Designed for use in labs for general cleaning, small spills, and hazardous spill containment
  • Large size: 12 in. x 13 in.
  • Heavy-duty, cloth-like material is low-lint, no loose fibers or threads
  • Available in 50-pack bags, 18 bags per case (900/case)

 

Specifications

Product Type Lab Wipes
Length (English) 12 in.
Length (Metric) 10.97 cm
Width (English) 13 in.
Width (Metric) 11.88 cm
Material Cloth
Color White
Dimensions (L x W) 12 x 13 in.
For Use With (Application) General cleaning, small spills, and hazardous spill containment