Description
Easy to clean, easy to cut.
Protects drawers, shelves, and benches from chemicals, spills, and dirt.
- Clear polyethylene ribbed matting cushions instruments, reduces glassware breakage
- Five times thicker (0.036 in.) than normal shelf liner, but won't ripple; lies flat
Chemical Identifiers
| Lab Mat | |
| 30 in. | |
| Clear | |
| 9 mm (0.036 in.) |
| Polyethylene | |
| 762 mm | |
| Protects drawers, shelves, and benches from chemicals, spills, and dirt | |
| 1 Ea. |
Specifications
| Lab Mat | |
| 15 ft. | |
| 30 in. | |
| 15 ft. x 30 in. (4.4 m x 76 cm) | |
| Protects drawers, shelves, and benches from chemicals, spills, and dirt | |
| 1 Ea. |
| Polyethylene | |
| 4.57 m | |
| 762 mm | |
| Clear | |
| 9 mm (0.036 in.) |