missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lab Hazard Warning Labels with Dispenser
GREENER_CHOICE

Catalog No. 15925
Change view
Click to view available options
Color:
Multi-colored
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15925 Multi-colored
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15925

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Printed in safety colors.

  • Dispenser supplied with four pressure-sensitive tape rolls preprinted with warnings in safety colors: POISON (yellow on black); CORROSIVE (black on yellow); FLAMMABLE (red on white); and RADIOACTIVE (red on yellow)
  • Serrated tear-off edge for easy tape separation
  • Approximately 200 imprints per roll
  • Refill rolls available separately

Order Info

Each tape roll measures 0.50 in.W x 22 yd. L (13mm x 20m)

Specifications

Color Multi-colored
Format Tape
Product Type Tape Dispenser
For Use With (Application) Printed in safety colors
Includes 4 Rolls with preprinted warnings corrosive (black on yellow), flammable (red on white), poison (yellow on black), radioactive (red on yellow)
Quantity 1 Ea.
Length (English) 22 yd.
Length (Metric) 20 m
Width (English) 0.5 in.