missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lab Hazard Warning Label Refills
GREENER_CHOICE

Catalog No. 15934
Change view
Click to view available options
Display Text:
Corrosive
Flammable
Poison
Radioactive
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
15934 Flammable
15931 Radioactive
15932 Poison
15933 Corrosive
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15934

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Printed in safety colors.

For use with Fisherbrand™ Lab Hazard Warning Label Dispenser (cat. no. 15931).

  • Tape rolls preprinted with warnings in safety colors: POISON (yellow on black); CORROSIVE (black on yellow); FLAMMABLE (red on white); and RADIOACTIVE (red on yellow)
  • Serrated tear-off edge for easy tape separation
  • Approximately 200 imprints per roll

Order Info

Each tape roll measures 0.50 in. W x 22 yd. L (13mm x 20m)

Specifications

Color Assorted
Format Tape
Product Type Flammable Warning Refill Tape
Display Text Flammable
For Use With (Application) Printed in safety colors
Quantity 4/Cs.
Length (English) 57 yd.
Length (Metric) 20.3 m
Width (English) 0.5 in.