missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lab Automation Pipet Tips

Catalog No. 02707217
Change view
Click to view available options
Volume (Metric):
1 to 20 μL
5 to 250 μL
20 μL
50 μL
100 μL
200 μL
250 μL
1100 μL
Sterility:
Non-sterile
Sterile
Color:
Black
Blue
Natural
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Color
02707217 250 μL Sterile Blue
02707216 250 μL Non-sterile Blue
02707236 1 to 20 μL Non-sterile Natural
02707237 1 to 20 μL Sterile Natural
02707239 5 to 250 μL Non-sterile Natural
02707240 5 to 250 μL Sterile Natural
02707283 1100 μL Non-sterile Natural
02707274 20 μL Sterile Black
02707341 100 μL Sterile Natural
02707342 200 μL Non-sterile Natural
02707344 50 μL Non-sterile Natural
02707343 200 μL Sterile Natural
02707345 50 μL Sterile Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 02707217

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

Carbon-black, liquid-level-sensing tips are required by many automated systems. They are constructed of a highly conductive material that is necessary to detect presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.

Disposable tips for robotic workstations — provide maximum performance, accuracy, and reproducibility in laboratory automation procedures. Available in clear, blue tint, or carbon-black-conductive polypropylene.
Features

  • Ideal for demanding HTS procedures
  • Provide maximum performance, accuracy, and reproducibility in lab automation procedures
  • Engineered to ensure proper fit and function with several standard and customized automated liquid-handling systems
  • Choose tips in an assortment of volume sizes and styles

Specifications

Volume (Metric) 250 μL
Sterility Sterile
Color Blue
Autoclavable Autoclavable
For Use With (Application) Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures
Format Rack
Tip Style Automation
Disposable Single-use
For Use With (Equipment) Beckman Coulter Multimek 96 Channel
Material Polypropylene