missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Inventory Label Tape
GREENER_CHOICE

Catalog No. 15907
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15907 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15907

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conveniently printed for inventory control needs.

  • Apply to bottles, boxes, vials—anything for which inventory control must be maintained
  • Each label provides space for notations after each of these printed headings: Date Received, Date Opened, Date Expired, and Reorder Number
  • Labels can be easily removed after application without leaving residue
  • White with dark blue imprint

Order Info

Labels measure 1W x 3 in. L (2.5 x 7.6cm). Use with 15-942 dispenser (sold separately).

Specifications

Color White
Format Tape
Product Type Inventory Label Tape
For Use With (Application) Used for bottles, boxes, vials and anything for which inventory control must be maintained
Quantity 12/Pk.
Length (English) 14 yd.
Length (Metric) 13 m
Width (English) 1 in.