Learn More
Com-Pac International™ Infecon™ Specimen Transport Bags
Document pouch keeps paperwork with the specimen.
Supplier:  Com-Pac International™ 5025IP
 View more versions of this product
Catalog No. 19075210
Only null left
Description
Description
Document pouch keeps paperwork with the specimen
  • For collecting, storing and transporting tissue samples
  • Patented seal and double-lock closure make bag liquid-tight, easy to open and close
  • Polyethylene with preprinted biohazard label
Specifications
Chemical Identifiers
Double-track Zip Seal
Polyethylene
Specifications
Double-track Zip Seal
16 x 16 in. (40.64 x 40.64cm)
16 in.
40.6 cm
Polyethylene
16 in.
40.6 cm
Product Suggestions
Product Suggestions