missing translation for 'onlineSavingsMsg'
Learn More

Com-Pac International™ Infecon™ Specimen Transport Bags

Catalog No. 18028321
Change view
Click to view available options
Dimensions (L x W):
10 x 8 in. (25.4 x 20.32cm)
12 x 12 in. (30.48 x 30.48cm)
12 x 9 in. (30.48 x 22.86cm)
16 x 16 in. (40.64 x 40.64cm)
9 x 6 in. (22.86 x 15.24cm)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18028321 10 x 8 in. (25.4 x 20.32cm)
19028320 12 x 12 in. (30.48 x 30.48cm)
19075210 16 x 16 in. (40.64 x 40.64cm)
19287215 9 x 6 in. (22.86 x 15.24cm)
22287216 12 x 9 in. (30.48 x 22.86cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18028321 Supplier Com-Pac International™ Supplier No. 5023IP

Document pouch keeps paperwork with the specimen.

Document pouch keeps paperwork with the specimen

  • For collecting, storing and transporting tissue samples
  • Patented seal and double-lock closure make bag liquid-tight, easy to open and close
  • Polyethylene with preprinted biohazard label

Specifications

Closure Type Double-track Zip Seal
Material Polyethylene
Dimensions (L x W) 10 x 8 in. (25.4 x 20.32cm)
Length (English) 10 in.
Width (English) 8 in.
Length (Metric) 25.4 cm
Width (Metric) 20.32 cm