missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Triple Layer Hot Mill Gloves - Band Top

Catalog No. 19013544
Change view
Click to view available options
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19013544 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19013544 Supplier Fisherbrand™ Supplier No. 94928I

Ideal for forging and other operations that require heat protection.

  • Constructed from three layers of cotton fabric
  • Burlap lining creates air pockets for increased heat protection
  • Ideal for forging and other operations that require heat protection

Specifications

Accelerator Free Yes
Applications Industrial
Coating Material Nap-out
Color Natural
Latex Free Yes
Material Cotton
Product Type Hot Mill Gloves
Size Universal
Style Clute
For Use With (Application) Forging, and Hot Metal Handling/Castings/Molds
Gender Men's
Glove Cuff Style Band Top
Liner Material Burlap
Series 94-928
Yarn Weight 28 oz.
Show More Show Less