Learn More
Fisherbrand™ Hollow Cathode Multi-Element 1 1/2in. Lamps
Catalog No. 14386103T
Change view
Click to view available options
Element:
Calcium, Magnesium, Zinc
Cr, Co, Cu, Fe, Mn, Ni
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Element
14386103T Cr, Co, Cu, Fe, Mn, Ni
14386104G Calcium, Magnesium, Zinc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14386103T
Window materials are quartz, borosilicate and UV glass; Lamps are multi-element and 1 1/2 in. (37mm) in diameter
  • Quartz windows with neon gas fill (sodium/potassium has UV glass)
  • Lamps are multi-element and have 1 1/2 in. diameter (37mm) bodies For use in all AA spectrophotometers except Perkin-Elmer™ instruments
  • May be adapted to fit PE™ instruments using the 14-386-102Z Adapter Kit (sold separately)
Specifications
Element Cr, Co, Cu, Fe, Mn, Ni
Gas Fill Neon
Light Source Hollow Cathode
Diameter (English) 1.5 in.
Diameter (Metric) 3.7 cm
For Use With (Equipment) Spectrophotometer
Material Quartz
Shape Y-Shape
Product Type Spectrophotometer Lamp