Learn More
Fisherbrand™ Histosette™ II Tissue and Biopsy Cassettes with Combination Lid and Base
Histosette™ II Tissue Processing/Embedding Cassettes with Separate Lid and Base open and close as often as necessary — cassettes will always relock securely.
Supplier:  Fisherbrand™ M4938
Catalog No. 15-182-702G
Only null left
Explore available promotions
Description
Description
  • Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
  • Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Specifications
Specifications
Slots
Combination Lid
Plastic
45°
Tan
Biopsy Cassette
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories