Learn More
Fisherbrand™ Histology Buckets
Flip-top lid helps to reduce noxious odors
$350.69 - $432.21
Specifications
Graduated No
Material Polypropylene
Closure Type Flip-top Snap-on Lid
View More Specs
Products 2
Catalog Number Capacity (Metric) Capacity (English) Price Quantity & Availability  
Catalog Number Capacity (Metric) Capacity (English) Price Quantity & Availability  
23111250
View Documents Promotion Details
2 L 0.5 gal.
Case of 20 Each for $432.21
Only null left
23111251
View Documents Promotion Details
5 L 1.37 gal.
Case of 10 Each for $350.69
Only null left
Description
Description
  • Opaque polypropylene construction resists cracking and chemicals, including formaldehyde
  • Metal carrying handle eases transport
  • Easy to stack and store
  • No graduation marks
  • Autoclavable
Specifications
Specifications
No
Polypropylene
Flip-top Snap-on Lid