missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ High Impact Polystyrene (HIPS) Drying Rack Pegs

Catalog No. 0571841
Change view
Click to view available options
Holds:
Labware
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
0571841 Labware
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0571841
Only null left

Polystyrene pegs for use with Fisherbrand™ Drying Rack.

Pegs for use with Fisherbrand™ Drying Rack.

  • Sturdy pegs hold cylinders, bottles, beakers and containers
  • Snap in or out easily when desired
  • Bag of 11 pegs made of high impact polystyrene

Specifications

Autoclavable Not Autoclavable
Color White
Holds Labware
Material Polystyrene
For Use With (Equipment) Fisherbrand™ High Impact Polystyrene (HIPS) Drying Rack (05-718-40)
Length (Metric) 12.42 cm
Length (English) 4.89 in.
Width (English) 0.576 in.
Product Type Pegs for Drying Rack
Width (Metric) 1.46 cm