Learn More
Fisherbrand™ Hemato-Seal™ Capillary Tube Sealant
Catalog No. 02678
Change view
Click to view available options
Quantity:
10 Pk.
Catalog No. Quantity
02678 10 Pk.
1 options
Catalog No. 02678
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Hemato-Seal™ Capillary Tube Sealant. Hemato-Seal™ Capillary Tube Sealant is a nondrying, nonhardening, nontoxic compound for sealing hematocrit and other blood collecting tubes. Its light color provides sharp contrast for easy reading and ensures excellent retention during centrifugation.
Nondrying, nonhardening, nontoxic compound for sealing hematocrit and other blood collecting tubes
  • Light color provides sharp contrast for easy reading
  • Excellent retention during centrifugation
Specifications
Material Clay
Product Type Capillary Tube Sealant
For Use With (Application) Sealing hematocrit and other Blood collecting tubes, provides excellent retention during centrifugation
For Use With (Equipment) Hemato-Seal™ capillary tube
Quantity 10 Pk.