Learn More
Fisherbrand™ Heavy-Duty Short-Blade Scissors
Fisherbrand Heavy-Duty Short-Blade Scissors have reinforced clip-point for cutting heavy fabric and paper
$28.62 - $36.28
Specifications
Product Type Heavy Duty Short-Blade Scissors
Blade Style Angled
Material Steel
For Use With (Application) Heavy Duty
View More Specs
Products 2
Catalog Number Length (Metric) Price Quantity & Availability  
Catalog Number Length (Metric) Price Quantity & Availability  
14275A
View Documents Promotion Details
15 cm
Each for $28.62
Only null left
14275B
View Documents Promotion Details
20.3 cm
Each for $36.28
Only null left
Description
Description
Fisherbrand Heavy-Duty Short-Blade Scissors have reinforced clip-point for cutting heavy fabric and paper
  • Fisherbrand™ Heavy-Duty Short-Blade Scissors perfect for various heavy-duty applications
  • Hot-hammer forged, hardened and tempered of special cutlery steel
  • Double-plated chrome over nickel with black enameled handles
  • Heavyweight, short-blade design
Specifications
Specifications
Heavy Duty Short-Blade Scissors
Angled
Steel
Heavy Duty