missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Heavyweight Cotton Gloves

Catalog No. 19048622A
Change view
Click to view available options
Accelerator Free:
Yes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Accelerator Free
19048622A Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19048622A Supplier Fisherbrand™ Supplier No. 35C410/L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Knit construction increases breathability and reduces hand fatigue.

  • Cotton/polyester seamless construction for added comfort
  • Reversible design allows replacing one glove at a time
  • Can be worn alone or used as a glove liner

Knit, Seamless Knit, Heavyweight, Cotton Gloves

Specifications

Construction String Knit
Accelerator Free Yes
Applications Industrial
Coating Material Uncoated
Color Natural
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Material Cotton
Powdered Powder Free
Product Type General Purpose Gloves
Size Large
Sterility Non-sterile
Glove Cuff Style Continuous Knit
Hem Color White
Yarn Gauge 7
Length (English) 10.6 in.
Length (Metric) 27 cm
Show More Show Less