missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Heavy-Paper Lined Sample Bags
Ideal for collection, shipment or storage of grain, seed, soil, or food samples
Description
Ideal for collection, shipment or storage of grain, seed, soil, or food samples
- Heavy-paper bags lined with 1mil-thick polyethylene
- Heat sealed at bottom to prevent seepage
- Tin-tie closure ensures moisture retention
Order Info
Maximum gusset width on side of bag is 2.5 in. (6.4cm) for 01-818-10; 3 in. (7.6cm) for 01-818-11.
Specifications
| Not Autoclavable | |
| Tin Tie | |
| Poly lined paper with tin tie | |
| 1000 | |
| 9.62 x 3.37 in. (24 x 8.6cm) | |
| 3.37 in. | |
| 2.5 in. |
| 1.5 lb. | |
| Assorted | |
| Paper | |
| 1 mil | |
| 24 cm | |
| 8.6 cm | |
| 6.4 cm |