missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Hazardous Chemical Dispensers
Bottle-top dispensers for use with hazardous and corrosive chemicals.
$280.93
Specifications
| Product Type | Chemical Bottle Pourer |
|---|---|
| Material | PTFE |
| Height (English) | 15 in. |
| Height (Metric) | 381 mm |
Description
- Constructed entirely of PTFE with totally inert pouring surfaces—even the neck-sealing ring is encapsulated in PTFE
- Incorporated breather vent allows for direct and controlled pourings
- Captured PTFE closure ensures contaminant-free contents when not in use
- Fit standard reagent bottles with neck sizes of GL32, 38, and 45
Specifications
| Chemical Bottle Pourer | |
| PTFE | |
| 15 in. | |
| 381 mm |