Learn More
Fisherbrand™ Graduated Cylinders
Single metric scale in tough ceramic enamel.
Supplier:  Fisherbrand™ S63457
 View more versions of this product
Catalog No. S63457
Only null left
Explore available promotions
Description
Description
  • Quality borosilicate glass cylinder with pouring lip
  • Heavy glass base provides extra stability
Specifications
Specifications
Round
Round
To Contain
50 mL
1 mL
±0.50 mL
±0.5 mL
Graduated Cylinder
Borosilicate Glass
Single
Autoclavable
1 mL
Product Suggestions
Product Suggestions