Learn More
Fisherbrand™ Kuehne Cover Glass Offset Thumb Forceps
For immersing cover glasses in preparation dish. Fisherbrand™ Kuehne Cover Glass Offset Thumb Forceps feature bent ends for grasping cover glasses.
Catalog No. 10-295
Only null left
Explore available promotions
Description
Description
  • Fisherbrand™ Kuehne Cover Glass Offset Thumb Forceps are available in bent or straight configuration. Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Broad, thin tips, bent at handy angle
  • Easy to reach into containers without obscuring the object being gripped
  • Grooved handles
  • Stainless-steel construction per ASTM™ specification
  • Can be sterilized by EtO/gamma radiation
  • Rockwell hardness (HRC): 40–45
  • Latex-free
Specifications
Specifications
Offset-tipped
FDA certified stainless-steel
Forceps
Thumb
4.37 in.
11.1 cm
Immersing cover glasses in preparation dish
Product Suggestions
Product Suggestions