missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Gel-Loading Aerosol-Barrier Tips
GREENER_CHOICE

Catalog No. p-159370
Change view
Click to view available options
Volume (Metric):
10 to 100 μL
10 μL
20 μL
Packaging:
Bulk
Racked
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Packaging
02707169 20 μL Bulk
02707171 20 μL Racked
02707174 10 to 100 μL Racked
02707175 10 μL Racked
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02707169
Only null left

Help minimize liquid retention

  • No RNase or DNase contamination
  • Natural color
  • For use with, but not limited to: Biohit Proline™ Electronic, BrandTech™ Transferpette™, Eppendorf™ Reference 2000, Eppendorf Research 2100, Thermo Finnpipette™ Focus, Fisherbrand™ Finnpipette™, Fisherbrand Finnpipette II, Genemate™ ISC Bio Express, Hamilton SoftGrip™, Jencons Sealpette™ Electronic, Labnet Labpette™ Digital, Mettler Toledo VoluMate™, Nichiryo 5000, Oxford™ Benchmate™, Pipetman™, Rainin™, Vistalab Ovation

Specifications

Volume (Metric) 20 μL
Filtered Filtered
Color Natural
Length (English) 2.7 in.
Length (Metric) 7cm
For Use With (Application) Help minimize liquid retention
Tip Style Gel Loading
Packaging Bulk
Material Polypropylene