missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Formalin Neutralizing/Absorbing Pads

Catalog No. p-3597427
Change view
Click to view available options
Color:
Blue
White
Packaging Quantity:
30 Pads/Cs.
55 Sheets/Roll, 6 Rolls/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Packaging Quantity
2202670 Blue 55 Sheets/Roll, 6 Rolls/Cs.
2202671 White 30 Pads/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2202670 Supplier Fisherbrand™ Supplier No. ES728F
Only null left

For absorbing and neutralizing formalin in a lab environment

  • Available in individual pad and handy roll formats for ease of use and economics
  • Treated for the ultimate destruction of 10% Neutral Buffered Formalin
  • Neutralizes and reduces harmful vapors during grossing procedures
  • Blue color of sheets provides excellent contrast for tissue grossing
  • Large individual pads absorb, encapsulate, and neutralize up to 900mL of 10% Neutral Buffered Formalin
  • Small absorbent pads are supplied on a roll

Small Absorbent Pads (Sheets on Rolls)

  • 11 × 10 in. sheets
  • Super Absorbent Pads:
    • 9 × 21 in. pads
    • Pads absorb, encapsulate, and neutralize up to 900cc of 10% Neutral Buffered Formalin

    Specifications

    Color Blue
    Type Formalin Absorbent Pad
    For Use With (Application) Neutralizes and reduces harmful vapors during grossing procedures
    Packaging Quantity 55 Sheets/Roll, 6 Rolls/Cs.