Learn More
Fisherbrand™ Flask Tongs
Catalog No. 10062
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
10062 Stainless Steel
1 options
Catalog No. 10062 Supplier Fisherbrand™ Supplier No. 29162
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Flasks can’t slip when gripped by these flask tongs. Fisherbrand™ Flask Tongs are made from ASTM™ standard stainless-steel.
Fisherbrand™ Flask Tongs. Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • For Erlenmeyer or Florence flasks taking Nos. 4 to 8 stoppers
  • All stainless steel
Specifications
Product Type Flask Tongs
Material Stainless Steel
For Use With (Application) Holding flasks