Learn More
Fisherbrand™ Erlenmeyer Flask Clamps for Ultrasonic Units
Safely and securely attach Erlenmeyer flasks to Fisherbrand™ ultrasonic cleaning units. Fisherbrand™ Erlenmeyer Flask Clamps are manufactured using strong and durable stainless steel for superior stability.
Supplier: Fisherbrand™ 1007557
Description
- Safe and secure attachment
- Manufactured from strong and durable stainless steel
- Provides superior stability
Chemical Identifiers
| Flask Clamp | |
| Ultrasonic Cleaner |
Specifications
| Flask Clamp | |
| Ultrasonic Cleaner | |
| 500 mL Erlenmeyer Flask |