Learn More
Fisherbrand™ Erlenmeyer Flask Clamps for Ultrasonic Units
Catalog No. 2940717
Change view
Click to view available options
For Use With (Application):
1000mL erlenmeyer flask
250mL Erlenmeyer flask
50 mL Erlenmeyer Flask
500 mL Erlenmeyer Flask
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
2940717 50 mL Erlenmeyer Flask
2940718 250mL Erlenmeyer flask
2940719 500 mL Erlenmeyer Flask
2940720 1000mL erlenmeyer flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 2940717 Supplier Fisherbrand™ Supplier No. 1007553
Only null left
Safely and securely attach Erlenmeyer flasks to Fisherbrand™ ultrasonic cleaning units. Fisherbrand™ Erlenmeyer Flask Clamps are manufactured using strong and durable stainless steel for superior stability.
  • Safe and secure attachment
  • Manufactured from strong and durable stainless steel
  • Provides superior stability
Specifications
Type Flask Clamp
For Use With (Equipment) Ultrasonic Cleaner
For Use With (Application) 50 mL Erlenmeyer Flask