Learn More
Fisherbrand™ End-to-End Connector
Catalog No. 02215477
Change view
Click to view available options
Material:
Aluminum Alloy
Catalog No. Material
02215477 Aluminum Alloy
1 options
Catalog No. 02215477 Supplier Fisherbrand™ Supplier No. 80862780
Strong aluminum alloy connector that permits end-to-end joining of rods up to 13mm (0.51 in.).
  • Extends length of lab frame rods
  • Precised boring of connector ensures perfect alignment of rods
  • Comes with two set of screws
  • Resistant to corrosion
Specifications
Type End-to-End Connector
Material Aluminum Alloy
For Use With (Application) For end-to-end joining of rods up to 0.51 in. (13mm)