missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Economy Grade Latex Coated Palm Gloves

Catalog No. 19121383
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19121383 Small
19816901 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19121383 Supplier Fisherbrand™ Supplier No. 39C121/S
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Economical palm-coated gloves combine cost with comfort for an effective level of protection.

  • Red latex, smooth palm coated
  • Coated palm offers abrasion resistance and increased grip
  • Uncoated back allows hands to breathe
  • Ideal for assembly, construction, inspection, material handling, refuse handling, and landscaping

Specifications

Coated Area Palm
Coating Material Latex
Color Natural
Grade Economy
Material Polycotton
Product Type Dip-coated Gloves
Size Small
For Use With (Application) Assembly, Construction, Inspection, Material Handling, and Landscaping
Glove Cuff Style Continuous Knit
Series 39-C121
Length (English) 8.5 in.
Length (Metric) 21.6 cm
Show More Show Less