missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micro Essential Lab Economical Plastic pH Strips
Description
- Packaged in zip lock pack
- Colorimetric chart located on the vial label
- Plastic pH test strip construction
- Ideal for lab, field or classroom use
- Instant dip and read result
Specifications
Specifications
| pH Range | 0 to 14 |
| Format | Plastic Strip |
| For Use With (Application) | For classroom experiments or demonstrations |
| Quantity | 50 Pk. |
| Storage Requirements | Room Temperature |
| Type | Plastic pH Test Strip |
This product is not intended to diagnose, treat, cure or prevent any disease or condition. Please contact a medical doctor to diagnose and treat any medical condition.