Learn More
Fisherbrand™ Dye Industry Pipets
Perfect for pipetting dark-color dye solutions.
 View more versions of this product
Catalog No. 13-671-109K
Only null left
Explore available promotions
Description
Description
Just fill pipet to desired volume and read—no time-consuming draining as with conventional pipets.
  • White graduations stand out against background of dark dye; easier to see and read than conventional amber markings
  • Inverse scale—volume graduations start with 0 at tip
  • Color coded for quick, accurate identification
  • Catalog number, capacity, and accuracy marked on each pipet
  • Tough borosilicate glass; Tuf-Temp™-treated to reduce chipping
Compliance
Calibrated To Deliver/Blow Out.
Specifications
Specifications
±0.04 mL
Borosilicate Glass
0.1 mL
Blue
Dye Industry Pipet
To Deliver
Yes
12/Cs.
5 mL
Product Suggestions
Product Suggestions