Learn More
Fisherbrand™ Dye Industry Pipets
Perfect for pipetting dark-color dye solutions.
Description
Just fill pipet to desired volume and read—no time-consuming draining as with conventional pipets.
- White graduations stand out against background of dark dye; easier to see and read than conventional amber markings
- Inverse scale—volume graduations start with 0 at tip
- Color coded for quick, accurate identification
- Catalog number, capacity, and accuracy marked on each pipet
- Tough borosilicate glass; Tuf-Temp™-treated to reduce chipping
Compliance
Calibrated To Deliver/Blow Out.
Specifications
| ±0.02 mL | |
| Yes | |
| 12/Cs. | |
| 1 mL |
| Borosilicate Glass | |
| 0.01 mL | |
| Yellow | |
| Dye Industry Pipet |