missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ DPD-Free Chlorine Test Reagent
Description
- Use with any DPD-based photometer
- Individual premeasured packages of reagent
Specifications
Specifications
| Volume (Metric) | 5 mL |
| For Use With (Application) | To test for chlorine in HF scientific and other manufacturer's photometers |
| For Use With (Equipment) | HF scientific and other manufacturer's photometers |
| Quantity | 200 Pack |
| Type | DPD Free Chlorine Test Reagent |
| Volume (English) | 0.16 oz. |