Learn More
Fisherbrand™ Cryogenic Dots in Laser Sheets
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Come in an easy to use dispenser box with 20 sheets per pack.
Description
The labels can be printed on and will not bleed. Perfect for distinguishing amongst various specimens.Chemical Identifiers
| Sheet | |
| Non-sterile | |
| 0.5 to 1.5mL tubes |
Specifications
| Yellow | |
| Dot Label | |
| 0.5 to 1.5mL tubes | |
| 9.55 mm |
| Sheet | |
| Non-sterile | |
| 0.37 in. |