Learn More
Fisherbrand™ Cryogenic Dots in Laser Sheets
GREENER_CHOICE
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Come in an easy to use dispenser box with 20 sheets per pack.
 View more versions of this product
Catalog No. 11010202
Only null left
Description
Description
The labels can be printed on and will not bleed. Perfect for distinguishing amongst various specimens.
Specifications
Chemical Identifiers
Sheet
Non-sterile
0.5 to 1.5mL tubes
Specifications
Green
Dot Label
0.5 to 1.5mL tubes
9.55 mm
Sheet
Non-sterile
0.37 in.
Product Suggestions
Product Suggestions