Learn More
Fisherbrand™ Dissecting Cushing Delicate Thumb Forceps
Catalog No. 08953C
Change view
Click to view available options
Tip Style:
Straight, serrated
Catalog No. Tip Style
08953C Straight, serrated
1 options
Catalog No. 08953C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fisherbrand Dissecting Cushing Delicate Thumb Forceps are made from ASTM™ stainless steel construction. These forceps are light/medium weight and are primarily used to handle delicate tissue.
  • Fisherbrand™ Dissecting Cushing Delicate Thumb Forceps are primarily used to handle delicate tissue
  • Our forceps are designed to exceed performance expectations -This is the Fisherbrand Advantage
  • Multipurpose - Useful in educational, research or healthcare setting
  • Delicate, straight tips with serrated jaws
  • Delicate- very easy to press together to avoid nerve pain with controlled handling
  • Patterned grip for easy and comfortable handling
  • Durable and resists rusting
  • Hand polished to mirror finish
  • Made from stainless steel
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully Autoclavable
  • Latex-free
  • Produced under ISO 9002, GMP, cGMP guidelines
Specifications
Product Type Dissecting Forcep
Autoclavable Autoclavable
Certifications/Compliance ASTM, FDA
Material Stainless Steel
Latex Free Yes
Tip Style Straight, serrated
Length (English) 7 in.
Length (Metric) 17.8 cm