Learn More
FISHERBRAND™ FISHERBRAND™ Disposable Wintrobe Tube
Catalog No. 22150730
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
22150730 1 Ea.
1 options
Catalog No. 22150730
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FISHERBRAND™ Disposable Wintrobe Tube is precision-scaled from the interior base and labelled with white enamel for easy reading. Ideal for determining sedimentation rates and hematocrit values in clinical and laboratory settings.
Labelled with white enamel for easy reading
  • Precision-scaled from the interior base for accurate readings
  • Labelled with white enamel for easy reading
  • Ideal for determining sedimentation rates and hematocrit values
  • Disposable design for single-use convenience
Specifications
Color Clear
Product Type Wintrobe Tube
Additive None
Material Soda Lime Glass
Sterility Non-sterile
Closure Type Tube Sealant
Quantity 1 Ea.