Learn More
FISHERBRAND™ FISHERBRAND™ Disposable Westergren Pipets
Catalog No. 22238444
Change view
Click to view available options
Quantity:
200/Pk.
Catalog No. Quantity
22238444 200/Pk.
1 options
Catalog No. 22238444
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FISHERBRAND™ Disposable Westergren Pipets are self-zeroing and designed to fit racks suitable for cotton-plugged Westergren pipets. Calibrated to assure the same normal values as conventional Westergren pipets for reliable ESR testing.
  • Designed to fit racks suitable for cotton-plugged Westergren pipets
  • Calibrated pipets assure the same normal values as conventional Westergren pipets
  • Self-zeroing design: when the cotton plug is completely saturated, cells automatically settle from the zero line
  • Disposable construction supports single-use workflows and reduces cross-contamination risk
Specifications
Color Clear
Product Type Westergren Tube
Additive None
Material Borosilicate Glass
Sterility Non-sterile
Closure Type Tube Sealant
Capacity (Metric) 1.5 mL
Quantity 200/Pk.
Volume (Metric) 1.5 mL