missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Graduated Transfer Pipettes
GREENER_CHOICE

Catalog No. p-163764
Change view
Click to view available options
Sterility:
Sterile
Bulb Draw (Metric):
3.1 to 3.2 mL
3.4 mL
Material:
LDPE
Polyethylene
Packaging Type:
10/Bag, 50 Bags/Pk.
20/Bag, 25 Bags/Pk.
Individually Wrapped, 500 Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Bulb Draw (Metric) Material Packaging Type
1371120 Sterile 3.1 to 3.2 mL LDPE Individually Wrapped, 500 Pk.
137119BM Sterile 3.4 mL Polyethylene 10/Bag, 50 Bags/Pk.
137119CM Sterile 3.4 mL Polyethylene 20/Bag, 25 Bags/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1371120
Only null left

Graduation marks allow multiple allocations in a single draw — useful anywhere there is a need for quick, safe transfer of fluids.

Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel. Molded in one piece from unbreakable, see-through low-density polyethylene to eliminate the hazards of broken glass and exposure to infectious materials.

  • 5.8 mL capacity with 0.25 mL graduations
  • Inert to biological fluids and most acids
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)

Order Info

Individually wrapped in peel-apart sleeve.

Specifications

Bulb Draw (Metric) 3.1 to 3.2 mL
Material LDPE
Sterility Sterile
For Use With (Application) Research
Graduated Yes
Graduations 0.25 mL
Quantity 500/Pk.
Packaging Type Individually Wrapped, 500 Pk.
Volume (Metric) 5.8 mL
Length (Metric) 15.4 cm
Type Transfer Pipet
Disposable Single-use
Show More Show Less